Anti-S1PR2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48126-50UL A48126-100UL
77714-121EA 228.38 USD
77714-121 77714-120
Anti-S1PR2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: S1PR2
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR