Anti-IMPDH1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A48095-50UG A48095-100UG
77714-068EA 228.38 USD
77714-068 77714-067
Anti-IMPDH1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: IMPDH1
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR