Anti-MMP8 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48014-50UL A48014-100UL
77713-922EA 228.38 USD
77713-922 77713-921
Anti-MMP8 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR