Anti-GK Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47720-100UL A47720-50UL
77713-426EA 334.95 USD
77713-426 77713-427
Anti-GK Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: GK
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR