Anti-GNRH1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47441-50UL A47441-100UL
77712-959EA 228.38 USD
77712-959 77712-958
Anti-GNRH1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: GNRH1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR