Anti-COL18A1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47440-100UL A47440-50UL
77712-956EA 334.95 USD
77712-956 77712-957
Anti-COL18A1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR