SM68 Standard 1: Al,As,B,Ba,Be,Bi,Ca,Cd,Ce,Co,Cr,Cs,Cu,Dy,Er,Eu,Fe,Ga,Gd,Ho,In,K,La,Li,Lu,Mg,Mn,Na,Nd,Ni,P,Pb,Pr,Rb,Re,Sc,Se,Sm,Sr,Tb,Th,Tl,Tm,U,V,Y,Yb,Zn @ 100 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-SM68-1-100 VHG-SM68-1-500
77697-861EA 452.15 USD
77697-861 77697-862
SM68 Standard 1: Al,As,B,Ba,Be,Bi,Ca,Cd,Ce,Co,Cr,Cs,Cu,Dy,Er,Eu,Fe,Ga,Gd,Ho,In,K,La,Li,Lu,Mg,Mn,Na,Nd,Ni,P,Pb,Pr,Rb,Re,Sc,Se,Sm,Sr,Tb,Th,Tl,Tm,U,V,Y,Yb,Zn @ 100 µg/mL in 5% HNO3, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards