Anti-RORA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47030-50UL A47030-100UL
77712-256EA 228.38 USD
77712-256 77712-255
Anti-RORA Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RORA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR