Anti-MSH2 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A46543-50UG A46543-100UG
77711-381EA 228.38 USD
77711-381 77711-380
Anti-MSH2 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR