Anti-CRISP3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60331-100UL A60331-50UL
77716-335EA 334.95 USD
77716-335 77716-336
Anti-CRISP3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: CRISP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR