Anti-HP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60239-100UL A60239-50UL
77716-184EA 334.95 USD
77716-184 77716-185
Anti-HP Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR