Anti-ST6GAL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48687-100UG A48687-50UG
77705-370EA 253.75 USD
77705-370 77715-106
Anti-ST6GAL1 Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-1:300, ELISA:1:5000-1:20000

Type:
Antigen: ST6GAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR