Anti-CLPTM1L Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64900-50UG A64900-100UG
77724-899EA 228.38 USD
77724-899 77724-898
Anti-CLPTM1L Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CLPTM1L
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR