Anti-OTUD5 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64863-100UG A64863-50UG
77724-829EA 334.95 USD
77724-829 77724-830
Anti-OTUD5 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OTUD5
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR