Anti-NEU4 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64697-50UG A64697-100UG
77724-511EA 228.38 USD
77724-511 77724-510
Anti-NEU4 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NEU4
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR