Anti-TTLL6 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61439-100UG A61439-50UG
77718-401EA 334.95 USD
77718-401 77718-402
Anti-TTLL6 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TTLL6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR