Anti-C7orf61 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A61134-100UG A61134-50UG
77717-832EA 334.95 USD
77717-832 77717-833
Anti-C7orf61 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: C7orf61
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR