Anti-CAMK1D Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61001-50UL A61001-100UL
77717-581EA 228.38 USD
77717-581 77717-580
Anti-CAMK1D Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: CAMK1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR