Anti-PGR Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A44515-100UG A44515-50UG
77707-784EA 334.95 USD
77707-784 77707-785
Anti-PGR Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PGR
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR