Anti-CSH1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44508-50UG A44508-100UG
77707-772EA 228.38 USD
77707-772 77707-771
Anti-CSH1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: CSH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR