Anti-STK36 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44044-100UL A44044-50UL
77706-970EA 334.95 USD
77706-970 77706-971
Anti-STK36 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:10-1:50

Type:
Antigen: STK36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR