Anti-DAZ4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61684-50UL A61684-100UL
77718-866EA 228.38 USD
77718-866 77718-865
Anti-DAZ4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:500-1:5000, IHC:1:5-1:20

Type:
Antigen: DAZ4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR