Anti-WDR24 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A62766-50UL A62766-100UL
77720-858EA 228.38 USD
77720-858 77720-857
Anti-WDR24 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: WDR24
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR