Multi-Element 11 Component Standard: Al @ 10µg/mL; Mg @ 20µg/mL; Ca @ 32µg/mL; Cu,Fe,Pb,Zn @ 50µg/mL; B @ 1000µg/mL; P @ 2000µg/mL; K @ 5000µg/mL; Na @ 10,000µg/mL in 3% HNO3 (v/v),VHG,LGC Standards
Supplier: LGC Standards
VHG-1201-100 VHG-1201-500
77692-988EA 208.6 USD
77692-988 77692-989
Multi-Element 11 Component Standard: Al @ 10µg/mL; Mg @ 20µg/mL; Ca @ 32µg/mL; Cu,Fe,Pb,Zn @ 50µg/mL; B @ 1000µg/mL; P @ 2000µg/mL; K @ 5000µg/mL; Na @ 10,000µg/mL in 3% HNO3 (v/v),VHG,LGC Standards
Standards Cationic Standards Multi-Element Standards