Sulfur Standard: S @ 100 µg/g (0.0100 wt%) in 20 cSt Mineral Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-S20MIN-100-100 VHG-S20MIN-100-500
77695-437EA 100.95 USD
77695-437 77695-438
Sulfur Standard: S @ 100 µg/g (0.0100 wt%) in 20 cSt Mineral Oil, VHG, LGC Standards
Standards Cationic Standards Sulphur Standards