Multi-Element Standard 23 Components: 1000 µg/mL each of Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Sr, Tl, Zn in 2% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-SM23-2-250 VHG-SM23-2-500
77693-258EA 329.55 USD
77693-258 77693-259
Multi-Element Standard 23 Components: 1000 µg/mL each of Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Sr, Tl, Zn in 2% HNO3, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards