Multi-Element Standard 21 components: 30 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-V21-30-100G VHG-V21-30-200G
77695-671EA 153.3 USD
77695-671 77695-672
Multi-Element Standard 21 components: 30 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards