Multi-Element Standard 23 components: 10 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-V23-10-100G
VHG-V23-10-200G
77695-691EA
203.75
USD
77695-691
77695-692
Multi-Element Standard 23 components: 10 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Standards
Cationic Standards
Multi-Element Standards