Aluminum Weighing Boats, Oil Free, Heathrow Scientific
Supplier: Heathrow Scientific
120932FR HEA14521AFR 120931FR 120935FR HEA14521BFR 120930FR HEA14521CFR 120936FR 120937FR
470357-472PK 35.9 USD
470357-472 470357-488 470357-478 470357-494 470357-496 470357-474 470357-476 470357-486 470357-484
Aluminum Weighing Boats, Oil Free, Heathrow Scientific
Boats Weighing Boats
Oil free boats for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
  • Processed and packaged in clean room environment
  • Lot traceable
  • Lightweight and sturdy
  • Able to withstand high temperatures
  • Resistant to many chemicals, including acids and bases
  • Finger tab handle can be etched for identification before weighing
  • Can be used for a wide range of applications including weighing, dispensing, mixing, and storing samples
  • Disposable
  • Available in a wide range of sizes
  • Sold in packs of 100
These Aluminum Weighing Boats areMore Product Information