NEX MD Fume Extractor Arms, Nederman
Supplier: Nederman
10561332 10561032 10561132 10561232
46102-036EA 4216.87 USD
46102-036 46102-030 46102-032 46102-034
NEX MD Fume Extractor Arms, Nederman
Fume Extractors
Ideal for medium- to heavy-duty applications and work environments with very heavy smoke, vapors, or non-explosive dust
Units feature ball joint hood suspension and parallelogram construction for easy and precise maneuverability of hood and arm. Robust swivel allows for 360° rotation. The encapsulated support mechanism protects the arm from dust and dirt. Hood design ensures high capture capacity and low noise level. Arms are equipped with 160mm (65/16") hose, which can be easily removed for cleaning. The hose, along with the external arm system, provides a high airflow and a low pressureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable