VACUUBRAND® PC3001 VARIO® pro Self-Adjusting Vacuum Pumping System, BrandTech
Supplier: BRANDTECH SCIENTIFIC INC.
20696703
97007-236EA 7857.26 USD
97007-236
VACUUBRAND® PC3001 VARIO® pro Self-Adjusting Vacuum Pumping System, BrandTech
Upgraded with over 25% more flowrate
The system is equipped with a CVC3000 vacuum controller with an intuitive menu-driven graphic interface that features a jog wheel for quick adjustments. The controller adjusts pump speed, providing smooth vacuum adjustment and eliminating the need for solenoid valves or pump switching. Offers control options previously unavailable with other vacuum pumps. Automatic evaporation control can find and follow boiling point curves, continuously optimizing vacuum levels for maximum evaporation speed without bumping. Single point vacuum control mode maintains stable, hysteresis-free vacuum at a single setpoint. Up to ten multi-step user programs can be entered for standardized operations.
The chemistry-design diaphragm pump eliminates the need to monitor and change oil. Pump flowpaths are constructed of corrosion-resistant fluoropolymers for minimal maintenance and a long service life. Solvent recovery accessories are constructed of premium, corrosion-resistant borosilicate glass. Pumps typically operate well in excess of 10,000 to 15,000 hours before servicing is required. A bi-directional RS-232 port is standard on all units.
Ideal vacuum system for benchtop rotary evaporation. System operates with minimal supervision, continuously optimizing the vacuum to changing application conditions for faster evaporation and tighter control. The system features a VARIO® version of the MD1C vacuum pump and integrated solvent recovery capabilities in a sturdy unit with a compact footprint. Additional benefits of the VARIO® control include significantly reduced noise and diaphragm wear.
Certifications: C/US marked. 230V model is CE marked.
Ordering information: Pump systems are supplied complete with CVC3000 controller, integral solvent recovery system, operating manual, and one-year manufacturer's warranty. The 230V model is supplied with a European-style plug.