Drum Cradle, Justrite®
Supplier: Justrite
08800 08308
56610-470EA 773.82 USD
56610-470 56610-620
Drum Cradle, Justrite®
Carts
The drum siphon and self-closing faucet provide a simple solution for nearly complete drainage of liquids out of 113.6 L (30 gal.) and 208.2 L (55 gal.) drums, eliminating the need for dangerous tipping or rolling.
  • Cradle allows easy transport of drums weighing up to 363 kg (800 lbs.)
  • Loading bar grips drum chime to ensure smooth movement between vertical and horizontal positions
  • Designed to roll right into Justrite drum cabinets
  • Four wheels
Unique siphon action empties residual solvents from drums, typically down to the last few ounces. Siphon and faucet are constructed of solid cast brass.
Certifications: Cradle is FM approved.