Gold ≥99.99%, disk, as rolled, Diameter 6.35 mm

Supplier: GOODFELLOW CORPORATION

1000030671 1000031920 1000030702 1000030568 1000031584 1000032086
77680-854EA 197.04 USD
77680-854 77681-135 77680-866 77680-825 77681-049 77681-179
Gold ≥99.99%, disk, as rolled, Diameter 6.35 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR