Gold ≥99.95%, disk, as rolled, Diameter 44.5 mm
Supplier: Goodfellow
1000044125
1000043627
1000046844
1000047067
77681-409EA
2811.61
USD
77681-409
77681-336
77681-773
77681-848
Gold ≥99.95%, disk, as rolled, Diameter 44.5 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...