Gold ≥99.95%, disk, as rolled, Diameter 5 mm

Supplier: Goodfellow

1000043974 1000043479 1000046643 1000046864
77681-341EA 196.56 USD
77681-341 77681-266 77681-705 77681-777
Gold ≥99.95%, disk, as rolled, Diameter 5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR