Gold ≥99.999%, disk, as rolled, Diameter 36 mm
Supplier: Goodfellow
1000019237
1000044979
1000046387
77680-716EA
3232.03
USD
77680-716
77681-549
77681-616
Gold ≥99.999%, disk, as rolled, Diameter 36 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...