Gold ≥99.9%, disk, Diameter 40 mm

Supplier: Goodfellow

1000030317 1000030418 1000030372 1000030524 1000031020 1000030468 1000031634 1000031744 1000031830 1000030814 1000031783
77680-740EA 558.45 USD
77680-740 77680-781 77680-766 77680-808 77680-934 77680-796 77681-059 77681-079 77681-104 77680-893 77681-090
Gold ≥99.9%, disk, Diameter 40 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR