Gold ≥99.9%, disk, Diameter 25 mm
Supplier: Goodfellow
1000030412
1000030370
1000030521
1000030314
1000030464
1000031629
1000031826
1000030808
77680-779EA
486.99
USD
77680-779
77680-764
77680-806
77680-738
77680-794
77681-058
77681-102
77680-891
Gold ≥99.9%, disk, Diameter 25 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...