ANR 94 ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A286839-100MG A286839-10MG A286839-25MG A286839-50MG A286839-5MG
ALA286839100MGEA 1209.26 USD
ALA286839100MG ALA286839-10MG ALA286839-25MG ALA286839-50MG ALA286839-5MG
ANR 94 ≥98%
ANR 94
Formula: C₉H₁₃N₅O
Storage Temperature: Ambient
CAS Number: 634924-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3974f4125913f0c70441a0973845a9e94a8f11f&vid=e3974f4125913f0c70441a0973845a9e94a8f11f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR