N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride ≥99%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Antalarmin hydrochloride, N-Butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride, N-butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride, Antalarmin HCl

A286834-10MG A286834-25MG A286834-50MG A286834-5MG
ALA286834-10MGEA 178.51 USD
ALA286834-10MG ALA286834-25MG ALA286834-50MG ALA286834-5MG
N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride ≥99%
N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride

Application:Antalarmin hydrochloride has been used:to study its effect on stress-induced tryptophan hydroxylase 2 activityto study the anxiogenic effects of corticotropin-releasing factor 1 (CRF1) activationto analyze the therapeutic effect of blocking corticotrophin-releasing hormone (CRH) signaling in treating social stress-induced depressionAntalarmin hydrochloride is a non-peptide CRF1 corticotropin-releasing factor receptor antagonist.

Formula: C₂₄H₃₅ClN₄
MW: 415.01 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05664721
CAS Number: 220953-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a51e71737bcadd6c9b5e373982d9abd6affd23d&vid=4a51e71737bcadd6c9b5e373982d9abd6affd23d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR