AMG 853 ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: 5-Chloro-4-[2-[[(2-chloro-4-cyclopropylphenyl)sulfonyl]amino]-4-[[(1,1-dimethylethyl)amino]carbonyl]phenoxy]-2-fluorobenzeneacetic acid, Vidupiprant

A286753-100MG A286753-10MG A286753-25MG A286753-50MG A286753-5MG
ALA286753100MGEA 1189.31 USD
ALA286753100MG ALA286753-10MG ALA286753-25MG ALA286753-50MG ALA286753-5MG
AMG 853 ≥98% (by HPLC), Moligand™
AMG 853
Formula: C₂₈H₂₇Cl₂FN₂O₆S
Storage Temperature: Freezer
CAS Number: 1169483-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52c874f3a9dd784eaab9cb96c29deeeafea746d3&vid=52c874f3a9dd784eaab9cb96c29deeeafea746d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR