(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide ≥99% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Tyrphostin A46, Tyrphostin 46, Tyrphostin AG-99, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide, AG 99, Tyrphostin 46,Tyrphostin A46,Tyrphostin B40, AG-99, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-2-propenamide, 2-Cyano-3-(3,4-dihydroxyphenyl)-2E-propenamide
A286649-50MG
A286649-100MG
A286649-10MG
ALA286649-50MGEA
112.01
USD
ALA286649-50MG
ALA286649100MG
ALA286649-10MG
(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide ≥99% (by HPLC)
(E)-2-Cyano-3-(3,4-dihydroxyphenyl)acrylamide
Formula:
C₁₀H₈N₂O₃ Melting Pt: 231 °C Storage Temperature: Freezer |
MDL Number:
MFCD00133901 CAS Number: 122520-85-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...