ATN-161 trifluoroacetate salt ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: (S)-2-((R)-2-((S)-2-((S)-2-((S)-1-Acetylpyrrolidine-2-carboxamido)-3-(1H-imidazol-4-yl)propanamido)-3-hydroxypropanamido)-3-mercaptopropanamido)succinamide TFA salt, ATN-161 TFA salt, ATN-161 Trifluoroacetate, PHSCN Peptide, Ac-PHSCN-NH2

A286606-10MG A286606-2MG A286606-50MG A286606-5MG A286606-100MG
ALA286606-10MGEA 215.56 USD
ALA286606-10MG ALA286606-2MG ALA286606-50MG ALA286606-5MG ALA286606100MG
ATN-161 trifluoroacetate salt ≥98%
ATN-161 trifluoroacetate salt
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eebc0ba4f9d62aea18f356ce4c3e08ecc0752ce&vid=1eebc0ba4f9d62aea18f356ce4c3e08ecc0752ce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR