Aluminium trilactate ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: L-lactate aluminum, Lactic acid aluminum salt, (±)-Lactic acid aluminium salt, L-lactate aluminum salt, Aluminum lactate, (±)-2-Hydroxypropanoic acid aluminium salt, Aluminium lactate, DL-Lactic acid aluminium salt

A283287-100G A283287-25G
ALA283287-100GEA 55.01 USD
ALA283287-100G ALA283287-25G
Aluminium trilactate ≥95%
Aluminium trilactate

It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.

Formula: (H₃CCH(OH)COO)₃Al
Melting Pt: >300 °C
MDL Number: MFCD20529175
CAS Number: 18917-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=273edf3a4bb0f61f89b766a2510c7716dd40ab34&vid=273edf3a4bb0f61f89b766a2510c7716dd40ab34&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR