rac-AntPhos ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-(Anthracen-9-yl)-3-(t-butyl-2,3-dihydrobenzo[d][1,3]oxaphosphole, 4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole, rac-AntPhos

A282056-250MG A282056-5G
ALA282056250MGEA 50.26 USD
ALA282056250MG ALA282056-5G
rac-AntPhos ≥97%
rac-AntPhos
Formula: C₂₅H₂₃OP
Storage Temperature: Refrigerator
MDL Number: MFCD30748199
CAS Number: 1268693-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d53443398b5643ca8b2e03ba4e79ae24d09cfbbe&vid=d53443398b5643ca8b2e03ba4e79ae24d09cfbbe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR