Methyl docosapentaenoate (7c,10c,13c,16c,19c) ≥99%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Methyl(7Z,10Z,13Z,16Z,19Z)-docosa-7,10,13,16,19-pentaenoate, all-cis-7,10,13,16,19-Methyl docosapentaenoate

A276308-100MG A276308-25MG
ALA276308100MGEA 504.36 USD
ALA276308100MG ALA276308-25MG
Methyl docosapentaenoate (7c,10c,13c,16c,19c) ≥99%
Methyl docosapentaenoate (7c,10c,13c,16c,19c)

Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₂₃H₃₆O₂
Storage Temperature: Ambient
MDL Number: MFCD01310506
CAS Number: 108698-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=501f71652691639ddcf2a1c160baed2b89696b6b&vid=501f71652691639ddcf2a1c160baed2b89696b6b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR