4-Azidobutyrate-N-hydroxysuccinimide ester ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (2,5-Dioxopyrrolidin-1-yl)4-azidobutanoate, Azidobutyric acid NHS ester, Succinimidyl 4-Azidobutanoate

A276210-10MG A276210-250MG A276210-25MG A276210-500MG A276210-50MG
ALA276210-10MGEA 31.26 USD
ALA276210-10MG ALA276210250MG ALA276210-25MG ALA276210500MG ALA276210-50MG
4-Azidobutyrate-N-hydroxysuccinimide ester ≥95%
4-Azidobutyrate-N-hydroxysuccinimide ester

Shipped at Room Temperature. Store at -20°C. Store under desiccating conditions.

Formula: C₈H₁₀N₄O₄
MW: 226.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22573606
CAS Number: 943858-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=321c75373bb73547554d740b91a7e53567ae04be&vid=321c75373bb73547554d740b91a7e53567ae04be&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR