AAL-993 ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: AAL 993, 2-[(4-Pyridyl)methyl]amino-N-[3-(trifluoromethyl)phenyl]benzamide, 2-[(4-Pyridinylmethyl)amino]-N-[3-(trifluoromethyl)phenyl]benzamide

A275890-100MG A275890-25MG A275890-50MG A275890-5MG
ALA275890100MGEA 2134.56 USD
ALA275890100MG ALA275890-25MG ALA275890-50MG ALA275890-5MG
AAL-993 ≥98%
AAL-993

Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₂₀H₁₆F₃N₃O
MW: 371.36 g/mol
Storage Temperature: Freezer
CAS Number: 269390-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb555515ef95f01c60c80389582dfc88b9f273e&vid=5cb555515ef95f01c60c80389582dfc88b9f273e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR