Anacardic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Hydroxy-6-pentadecylbenzoic acid

A275817-100MG A275817-10MG A275817-25MG A275817-50MG A275817-5MG
ALA275817100MGEA 535.71 USD
ALA275817100MG ALA275817-10MG ALA275817-25MG ALA275817-50MG ALA275817-5MG
Anacardic acid ≥98%
Anacardic acid

Store at -20°C. It is important to note that this product is reported to be light sensitive. Store In the Dark. Store under desiccating conditions.

Formula: C₂₂H₃₆O₃
MW: 348.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368254
CAS Number: 16611-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9eae8dc7760ee11b78fa3870e2cfe0e8f006b5ad&vid=9eae8dc7760ee11b78fa3870e2cfe0e8f006b5ad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR