Acacetin ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin
A275799-100MG
A275799-250MG
A275799-25MG
ALA275799100MGEA
310.56
USD
ALA275799100MG
ALA275799250MG
ALA275799-25MG
Acacetin ≥98%
Acacetin
Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.
Formula:
C₁₆H₁₂O₅ MW: 284.27 g/mol Melting Pt: 269 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016936 CAS Number: 480-44-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...